missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Microlink Caps
Description
Open top caps are ideal for storing and containing volatile solvents, sensitive compounds and corrosive chemicals
- Polypropylene/silicone/PTFE liner is ultrasonically welded to each cap to help eliminate possibility of toxicity
- Autoclavable
Specifications
Specifications
| Color | Black |
| Material | Polypropylene |
| No. per Case | 50/Cs. |
| Liner Material | Polypropylene/Silicone/PTFE |
| Neck Finish | 33-430 |
| For Use With (Equipment) | Bottle |
| Autoclavable | Autoclavable |
| Certifications/Compliance | ISO 9001 |
| Product Type | Open Top Screw Cap |