missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Media/Laboratory Bottle Caps
For use with Wheaton media bottles
$404.33 - $544.42
Specifications
| Color | Black |
|---|---|
| No. per Case | 200 Cs. |
| For Use With (Equipment) | Media bottle |
| Catalog Number | Mfr. No. | Neck Finish | Material | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Neck Finish | Material | Price | Quantity | ||||
|
06-406-31
|
DWK Life Sciences
240280 |
33-430 | Rubber |
Case of 200 Each for $417.62
|
|
||||
|
06-450-7
|
DWK Life Sciences
240080 |
33-430 | Polyethylene |
Pack of 200 Each for $404.33
|
|
||||
|
06-450-35
|
DWK Life Sciences
240281 |
38-430 | Rubber |
Pack of 200 Each for $544.42
|
|
||||
|
06-450-8
|
DWK Life Sciences
240081 |
38-430 | Polyethylene |
Pack of 200 Each for $506.40
|
|
||||
Description
- Black phenolic caps with screw thread
- Low Density Polyethylene (Not autoclavable), Rubber lined (Autoclavable)
Specifications
| Black | |
| 200 Cs. | |
| Media bottle |