Learn More
DWK Life Sciences Wheaton™ Low-Density Extended Controlled Dropper Tips (15mm)
15mm extended controlled dropper tip made of LDPE (Low Density Polyethylene)
$471.08 - $471.08
Specifications
| Certifications/Compliance | ISO 9001 |
|---|---|
| Drop Size | 41 μL |
| Product Type | Dropper Tip |
| For Use With (Equipment) | Plastic dropping bottle |
Description
- Extended controlled dropper tip dispenses one drop at a time
- Average drop size is 44μL ±6μL
Specifications
| ISO 9001 | |
| 41 μL | |
| Dropper Tip | |
| Plastic dropping bottle |