missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Low-Density Extended Controlled Dropper Tips
Description
- Extended controlled dropper tip dispenses one drop at a time
- Average drop size is 44μL ±6μL
- Case of 1000
Specifications
Specifications
| Color | Natural |
| Diameter (Metric) Inner | 8 mm |
| Material | LDPE |
| No. per Case | 1000 Cs. |
| Certifications/Compliance | ISO 9001 |
| Drop Size | 41 μL |
| Shape | Round |
| Product Type | Dropper Tip |
| For Use With (Equipment) | Plastic dropping bottle |