missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Low-Density Extended Controlled Dropper Tips (13mm)

Catalog No. 0344058
Change view
Click to view available options
Color:
Natural
White
Material:
Polyethylene
No. per Case:
1000 Cs.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Material No. per Case
0344058 Natural Polyethylene 1000 Cs.
0344048 White Polyethylene 1000 Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 0344058 Supplier DWK Life Sciences Supplier No. W24241101A

13mm extended controlled dropper tip made of LDPE (Low Density Polyethylene)

  • Extended controlled dropper tip dispenses one drop at a time
  • Average drop size is 44μL ±6μL
  • Case of 1000

Specifications

Color Natural
Material Polyethylene
No. per Case 1000 Cs.
Drop Size 41 μL
For Use With (Equipment) Plastic dropping bottle
Certifications/Compliance ISO 9001
Product Type Dropper Tip