missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences WHEATON™ Hybridization Rack

Catalog No. 1324598
Change view
Click to view available options
Material:
Acrylic
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
1324598 Acrylic
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 1324598 Supplier DWK Life Sciences Supplier No. DWK805015

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The WHEATON™ Hybridization Rack is an acrylic rack that holds up to six hybridization bottles of different sizes, providing safe and convenient transport or storage in the laboratory.

  • Acrylic rack holds up to six hybridization bottles of different sizes
  • Suitable for safe transport or storage of hybridization bottles
  • Compatible with various hybridization bottle sizes for flexible laboratory use

Order Info

Items come as one per case

Specifications

Product Type Hybridization Bottle
Diameter (Metric) Inner 35 mm
Diameter (English) Inner 1.37 in.
Material Acrylic
Coating Plastisol
Certifications/Compliance USP Type I
For Use With (Equipment) Hybridization bottle
For Use With (Application) Nucleic acid and protein blotting procedures that require a minimum of probe solution
Color Clear