Learn More
DWK Life Sciences Wheaton™ Headspace Standard Seals
Catalog No. 03410725
Change view
Click to view available options
Septum:
PTFE, Butyl
PTFE, Gray Butyl Molded
PTFE, Silicone
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Septum
03410725 PTFE, Butyl
03410724 PTFE, Silicone
03410726 PTFE, Gray Butyl Molded
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 03410725 Supplier DWK Life Sciences Supplier No. W224224
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Pressure release seals feature score lines that allow for internal pressure release when 3.0 ± 0.5 bar has been exceeded
  • Seals are made from aluminum for easy crimping and decapping
Specifications
Closure Color Natural
Septum PTFE, Butyl
Closure Material Aluminum
For Use With (Equipment) Crimp Top Headspace Vials
Product Type Headspace Crimp Seal