missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Headspace Seals with Pressure Release

Catalog No. 12141611
Change view
Click to view available options
Color:
Natural
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
12141611 Natural
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 12141611 Supplier DWK Life Sciences Supplier No. W224217

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Headspace seals feature aluminum‐faced silicone septa with pressure‐release and magnetic options for secure autosampler compatibility.

  • Offered in Standard, Pressure Release and Magnetic seals
  • Pressure release seals feature score lines that allow for internal pressure release when 3.0 ± 0.5 bar has been exceeded
  • Magnetic seals allow for use in magnetic transport autosamplers
  • Magnetic Seals are made from steel for easy crimping and decapping

Specifications

Classification Seals
Color Natural
Diameter (Metric) Outer 20.7 mm
Material Aluminum Faced Silicone
Pre-slit No
Sterility Non-sterile
Disposable Single-use
For Use With (Application) Headspace vial sealing for GC analysis
Style Pressure release
Product Line Wheaton
Dnase Free No
Quantity 100
For Use With (Equipment) Crimp‐top headspace vials and GC autosamplers
Fit for Purpose Certified Quality
Autoclavable Not Autoclavable
Product Type Headspace Seal
Show More Show Less