missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Green Starline Polypropylene Caps for Redi-Pak™ Narrow-Mouth Containers

Catalog No. 0987710R
Change view
Click to view available options
Neck Finish:
13-425
20-410
No. per Case:
72 Cs.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish No. per Case
0987710R 13-425 72 Cs.
0987711E 20-410 72 Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 0987710R Supplier DWK Life Sciences Supplier No. 23950106

Caps that are the perfect fit for WHEATON Leak Resistant Narrow Mouth Bottles

  • Leak resistant
  • Autoclavable

Specifications

Color Green
Material Polypropylene
No. per Case 72 Cs.
Neck Finish 13-425
Certifications/Compliance ISO 9001
Product Type Polypropylene Cap
For Use With (Equipment) Redi-pak™ leak resistant narrow mouth bottle