Learn More
DWK Life Sciences Wheaton™ Glass Culture Vials
Culture vials feature deep-skirted screw caps to allow safer, more dependable handling when working with infectious materials
Supplier:  DWK Life Sciences 225142
 View more versions of this product
Catalog No. 06-451-190
Only null left
Explore available promotions
Description
Description
  • Wheaton 33 low extractable borosilicate glass
  • Deep-skirted black phenolic screw cap with 14B rubber liner
  • Autoclavable
Specifications
Specifications
14B Rubber
15 mm
4 mL
1.08 dr.
Screw Cap
ASTM E438, USP Type I, Type I, Class A
Clear
Borosilicate Glass
Screw
Phenolic
Autoclavable
49 mm
Product Suggestions
Product Suggestions