Learn More
DWK Life Sciences Wheaton™ E-C Sample Vials
Economically priced by packaging vials and screw caps separately. Wheaton™ E-C Sample Vials are available in either clear or amber-colored glass.
Supplier:  DWK Life Sciences 224751
 View more versions of this product
Catalog No. 033437B
Only null left
Description
Description
Economically priced by packaging vials and screw caps separately. Wheaton™ E-C Sample Vials are available in either clear or amber-colored glass.
  • Clear vials made from low extractable borosilicate glass that provides superior chemical resistance
  • Clear vials conform to USP Type I and ASTM E 438 Type I, Class A requirements
  • Amber vials made from borosilicate glass that conforms to USP Type I requirements for light transmission to protect light-sensitive products
  • Vials packed in shrink-wrapped packs of 100 which reduces corrugate contamination
  • 14B rubber lined black phenolic screw caps for aqueous samples
  • PTFE-faced 14B rubber lined black phenolic screw cap for organic samples
  • Vials and caps are autoclavable
Specifications
Chemical Identifiers
PTFE/14B Rubber
Screw
Screw Cap
ASTM Type I, USP Type I, Class A
Borosilicate Glass
Phenolic
Autoclavable
Specifications
PTFE/14B Rubber
12 mm
2.0 mL
0.5 dr.
Screw Cap
Autoclavable
38 mm
Amber
Borosilicate Glass
Screw
Phenolic
8-425
ASTM Type I, USP Type I, Class A
Product Suggestions
Product Suggestions