Learn More
DWK Life Sciences DURAN™ PURE Premium Screw Caps
Catalog No. 13710921
Change view
Click to view available options
Thread Size (Metric):
GL 25
GL 45
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Thread Size (Metric)
13710921 GL 25
13710922 GL 45
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13710921 Supplier DWK Life Sciences Supplier No. 891081306
Suitable for most sterilization methods
  • Made from fluoropolymer
  • Material contact surfacesmade from PTFE
  • Colorant and pigment free
  • Available in two DIN Thread sizes: GL 25 and GL 45
Specifications
Description Premium Screw Cap, DIN Thread Size: GL 25
Material Fluoropolymer
Thread Size (Metric) GL 25