missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Deck Kit for Compact Roller System for Small Bottles

Catalog No. 22288533
Change view
Click to view available options
For Use With (Equipment):
Compact small bottle roller culture apparatus
Wheaton benchtop small roller culture apparatus system
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
22288533 Wheaton benchtop small roller culture apparatus system
0644029 Compact small bottle roller culture apparatus
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22288533 Supplier DWK Life Sciences Supplier No. W348930CH

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Each deck holds two roller bottles

  • Attaches quickly and easily to Wheaton Compact Roller System for Small Bottles (above), which will accept two additional decks
  • Kit includes assembled roller deck, four support posts, drive belt, necessary hardware and assembly instructions

Order Info

Weight: 5 lb. (2.3kg)

Specifications

Product Type Deck Kit
Color Charcoal
For Use With (Equipment) Wheaton benchtop small roller culture apparatus system
Includes Assembled roller deck, 4 support posts, drive belt, necessary hardware and assembly instructions