missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences WHEATON™ COMPLETEPAK™ Sterile Components Vials

Catalog No. 15112170A
Change view
Click to view available options
Color:
Amber
Clear
Volume (Metric):
2 mL
5 mL
10 mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Volume (Metric)
15112170A Amber 10 mL
15112168A Amber 2 mL
15112169A Amber 5 mL
06451238A Clear 2 mL
06451239A Clear 5 mL
06451240A Clear 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 15112170A Supplier DWK Life Sciences Supplier No. W987002AS

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The WHEATON™ COMPLETEPAK™ product line is a range of crimp top vials, rubber stoppers and seals that are sterilized ready-to-use and are off- the-shelf, and can be customized for any application reducing your supply chain due to our one source solution capability.

  • Sterilized, off-the-shelf, ready-to-use (RtU) products
  • Variety of Kitted and stand-alone sterile components – a solution for any application
  • Complete manufacturing & certificate of analysis traceability
  • Ready-to-Use, Just-in-Time, minimum 3 month shelf life from time of shipment
  • Amber versions for light sensitive content packaging
  • Certifications: USP <788>/EP 2.9.19 Particulate Matter in Injections, USP <85>/EP 2.6.1, The Bacterial Endotoxins Test and lot certified to SAL <10-6, USP <660>/EP 3.2.1 Glass Containers for Pharmaceutical Use, and USP <381>/EP 3.2.9 Elastomeric Closures used in Parenteral Preparations

Specifications

Color Amber
Material Borosilicate Glass
Pre-slit No
Sterility Sterile
Volume (Metric) 10 mL
For Use With (Application) Light Sensitive Parenteral Preparations and Ready-to-Use Applications
Graduated No
No. per Case 104/Cs.
Style Crimp top
Product Line Wheaton CompletePAK
Marking Spot No
Autoclavable Autoclavable
Height (Metric) 50 mm
Product Type Sterile Vial
Show More Show Less