Learn More
DWK Life Sciences WHEATON™ CompletePAK™ Sterile Components Vials
The WHEATON™ CompletePAK™ product line is a range of crimp top vials, rubber stoppers and seals that are sterilized ready-to-use and are off- the-shelf, and can be customized for any application reducing your supply chain due to our one source solution capability.
Supplier:  DWK Life Sciences W987001AS
 View more versions of this product
Catalog No. 15112169A
Only null left
Description
Description
  • Sterilized, off-the-shelf, ready-to-use (RtU) products
  • Variety of Kitted and stand-alone sterile components – a solution for any application
  • Complete manufacturing & certificate of analysis traceability
  • Ready-to-Use, Just-in-Time, minimum 3 month shelf life from time of shipment
  • Amber versions for light sensitive content packaging
Certifications
USP <788>/EP 2.9.19 Particulate Matter in Injections, USP <85>/EP 2.6.1, The Bacterial Endotoxins Test and lot certified to SAL <10-6, USP <660>/EP 3.2.1 Glass Containers for Pharmaceutical Use, and USP <381>/EP 3.2.9 Elastomeric Closures used in Parenteral Preparations
Specifications
Chemical Identifiers
Sterile
Wheaton CompletePAK
Specifications
Amber
No
5 mL
No
Crimp top
No
Borosilicate Glass
Sterile
Light Sensitive Parenteral Preparations and Ready-to-Use Applications
120/Cs.
Wheaton CompletePAK
40 mm
Product Suggestions
Product Suggestions