Learn More
DWK Life Sciences WHEATON™ COMPLETEPAK™ Sterile Components Stoppers
Catalog No. 06451243A
Change view
Click to view available options
Closure Size:
13 mm
20 mm
Style:
OmniFlex 3G Igloo
OmniFlex 3G Straight Plug
Ultra Pure Straight Plug
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Size Style
06451243A 13 mm OmniFlex 3G Igloo
06451241A 13 mm OmniFlex 3G Straight Plug
06451242A 13 mm Ultra Pure Straight Plug
06451246A 20 mm OmniFlex 3G Igloo
06451244A 20 mm OmniFlex 3G Straight Plug
06451245A 20 mm Ultra Pure Straight Plug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 06451243A Supplier DWK Life Sciences Supplier No. W988005S
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
The WHEATON™ COMPLETEPAK™ product line is a range of crimp top vials, rubber stoppers and seals that are sterilized ready-to-use and are off- the-shelf, and can be customized for any application reducing your supply chain due to our one source solution capability.
  • Sterile
  • Stopper
Certifications
USP <788>, <85> and <71>; Container Closure Integrity (CCI) certified
Specifications
Material Bromobutyl
Sterility Sterile
Disposable Single-use
No. per Case 285/Cs.
Style OmniFlex 3G Igloo
Product Line Wheaton CompletePAK
Closure Size 13 mm
Autoclavable Autoclavable
Certifications/Compliance USP 788, 85, and 71, Container Closure Integrity (CCI) certified
Product Type Sterile Stopper