Learn More
DWK Life Sciences WHEATON™ CompletePAK™ Sterile Components Seals
The WHEATON™ CompletePAK™ product line is a range of crimp top vials, rubber stoppers and seals that are sterilized ready-to-use and are off- the-shelf, and can be customized for any application reducing your supply chain due to our one source solution capability.
Supplier:  DWK Life Sciences W988009S
 View more versions of this product
Catalog No. 06451247A
Only null left
Description
Description
  • Sterile
  • Flip-off Seal
Certifications
USP <788>, <85> and <71>; Container Closure Integrity (CCI) certified
Specifications
Chemical Identifiers
Aluminum
Wheaton CompletePAK
USP 788, 85, and 71, Container Closure Integrity (CCI) certified
Sterile
Sterile Flip Seal
Specifications
Red
Sterile
Wheaton CompletePAK
13 mm
USP 788, 85, and 71, Container Closure Integrity (CCI) certified
Aluminum
285/Cs.
Sterile Flip Seal
2 mL Serum Vial
Product Suggestions
Product Suggestions