missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences WHEATON™ COMPLETEPAK MODPAK Amber Kit

Catalog No. 15112215
Change view
Click to view available options
Diameter (Metric) Outer:
22 mm
24 mm
Closure Color:
Blue
Red
Closure Type:
Omni Flex Igloo Lyo Stopper
Omni Flex Serum Stopper
Ultra Pure Serum Stopper
Quantity:
104/Pk.
278/Pk.
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Outer Closure Color Closure Type Quantity
15112215 22 mm Blue Omni Flex Igloo Lyo Stopper -
15112207 - Blue Ultra Pure Serum Stopper 278/Pk.
15112208 - Blue Omni Flex Serum Stopper 278/Pk.
15112209 - Blue Omni Flex Igloo Lyo Stopper 278/Pk.
15112210 - Red Ultra Pure Serum Stopper 278/Pk.
15112211 - Red Omni Flex Serum Stopper 278/Pk.
15112212 - Red Omni Flex Igloo Lyo Stopper 278/Pk.
15112213 22 mm Blue Ultra Pure Serum Stopper -
15112214 22 mm Blue Omni Flex Serum Stopper -
15112216 22 mm Red Ultra Pure Serum Stopper -
15112217 22 mm Red Omni Flex Serum Stopper -
15112218 22 mm Red Omni Flex Igloo Lyo Stopper -
15112219 24 mm Blue Ultra Pure Serum Stopper 104/Pk.
15112220 24 mm Blue Omni Flex Serum Stopper 104/Pk.
15112221 24 mm Blue Omni Flex Igloo Lyo Stopper 104/Pk.
15112222 24 mm Red Ultra Pure Serum Stopper 104/Pk.
15112223 24 mm Red Omni Flex Serum Stopper 104/Pk.
15112224 24 mm Red Omni Flex Igloo Lyo Stopper 104/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
Catalog No. 15112215 Supplier DWK Life Sciences Supplier No. W991008ACPMP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Range of crimp top vials, rubber stoppers and seals that are sterilized ready-to-use and are off- the-shelf, and can be customized for any application reducing your supply chain due to our one source solution capability.

  • Six inventory matched kits conveniently packed into a single shipper case.
  • Sterilized, off-the-shelf, ready-to-use (RtU) primary packaging components
  • Focuses your effort on core competencies and reduces lead time
  • Streamlines process and frees up valuable clean room space USP /EP 2.9.19
  • Particulate Matter in Injections, USP /EP 2.6.1,
  • The Bacterial Endotoxins Test and USP / EP 2.6.14
  • The Sterility Test, USP /EP 3.2.1
  • Glass Containers for Pharmaceutical Use
  • USP /EP 3.2.9 Elastomeric Closures used in Parenteral Preparations
  • Complete manufacturing & certificate of analysis traceability
  • Ready-to-Use, Just-in-Time
  • Minimum 180 day shelf life from time of shipment
  • Container Closure Integrity (CCI) validation
  • Available in amber for photosensitive contents

Specifications

Closure Color Blue
Color Amber
Diameter (Metric) Outer 22 mm
Material Borosilicate Glass
Sterility Sterile
Disposable Single-use
Volume (Metric) 5 mL
For Use With (Application) Compounding pharmaceuticals, Aseptic filling of injectable drug products, Drug discovery, Drug research, Freeze drying / lyophilized drug products, Large and small molecule drug packaging
Graduated No
Includes Vial, Stopper, and Seal
Product Line CompletePAK ModPAK
Closure Material Bromobutyl
Closure Type Omni Flex Igloo Lyo Stopper
Closure Size 20 mm
Autoclavable Autoclavable
Certifications/Compliance USP <788>/EP 2.9.19 Particulate Matter in Injections, USP <85>/EP 2.6.1, The Bacterial Endotoxins Test and USP <71>/ EP 2.6.14 The Sterility Test, USP <660>/EP 3.2.1 Glass Containers for Pharmaceutical Use, and USP <381>/EP 3.2.9 Elastomeric Closures used in Parenteral Preparations
Height (Metric) 40 mm
Product Type Serum Vial
Show More Show Less