missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences WHEATON™ COMPLETEPAK™ Clear Kits

Catalog No. 06451226A
Change view
Click to view available options
Volume (Metric):
2 mL
5 mL
10 mL
Product Line:
OmniFlex 3G Igloo Lyo
OmniFlex 3G Serum
Ultra Pure Serum
Quantity:
104/Cs.
120/Cs.
278/Cs.
Closure Color:
Blue
Red
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Product Line Quantity Closure Color
06451226A 5 mL Ultra Pure Serum 120/Cs. Blue
06451222A 2 mL OmniFlex 3G Igloo Lyo 278/Cs. Blue
06451225A 2 mL OmniFlex 3G Igloo Lyo 278/Cs. Red
06451221A 2 mL OmniFlex 3G Serum 278/Cs. Blue
06451224A 2 mL OmniFlex 3G Serum 278/Cs. Red
06451220A 2 mL Ultra Pure Serum 278/Cs. Blue
06451223A 2 mL Ultra Pure Serum 278/Cs. Red
06451228A 5 mL OmniFlex 3G Igloo Lyo 120/Cs. Blue
06451231A 5 mL OmniFlex 3G Igloo Lyo 120/Cs. Red
06451227A 5 mL OmniFlex 3G Serum 120/Cs. Blue
06451230A 5 mL OmniFlex 3G Serum 120/Cs. Red
06451229A 5 mL Ultra Pure Serum 120/Cs. Red
06451234A 10 mL OmniFlex 3G Igloo Lyo 104/Cs. Blue
06451237A 10 mL OmniFlex 3G Igloo Lyo 104/Cs. Red
06451233A 10 mL OmniFlex 3G Serum 104/Cs. Blue
06451236A 10 mL OmniFlex 3G Serum 104/Cs. Red
06451232A 10 mL Ultra Pure Serum 104/Cs. Blue
06451235A 10 mL Ultra Pure Serum 104/Cs. Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
Catalog No. 06451226A Supplier DWK Life Sciences Supplier No. W991006CP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The WHEATON™ COMPLETEPAK™ product line is a range of crimp top vials, rubber stoppers and seals that are sterilized ready-to-use and are off- the-shelf, and can be customized for any application reducing your supply chain due to our one source solution capability.

  • Sterilized, off-the-shelf, ready-to-use (RtU) products
  • Variety of Kitted and stand-alone sterile components – a solution for any application
  • Kitted solutions feature equal amounts of RtU components for reduced scrap rates
  • Complete manufacturing & certificate of analysis traceability
  • USP 788/EP 2.9.19 Particulate Matter in Injections, USP 85/EP 2.6.1, The Bacterial Endotoxins Test and USP 71/ EP 2.6.14 The Sterility Test, USP 660/EP 3.2.1 Glass Containers for Pharmaceutical Use, and USP 381/EP 3.2.9 Elastomeric Closures used in Parenteral Preparations
  • CompletePAK™ ModPAK Bulk configurations
  • Ready-to-Use, Just-in-Time, minimum 3 month shelf life from time of shipment
  • Container Closure Integrity (CCI) certification available upon request

Certifications

USP <788>, <85> and <71>; Container Closure Integrity (CCI) certified

Specifications

Closure Color Blue
Material Glass
Sterility Sterile
Volume (Metric) 5 mL
Includes Vial, Stopper, and Seal
No. per Case 120/Cs.
Product Line Ultra Pure Serum
Closure Type Ultra Pure Serum Stopper
Quantity 120/Cs.
Closure Size 20 mm
Autoclavable Autoclavable
Certifications/Compliance USP 788, 85, and 71, Container Closure Integrity (CCI) certified
Product Type COMPLETEPAK Clear
Show More Show Less