Learn More
DWK Life Sciences WHEATON™ CompletePAK™ Clear Kits
Catalog No. 06451222A
Change view
Click to view available options
Volume (Metric):
2 mL
5 mL
10 mL
Closure Type:
OmniFlex 3G
OmniFlex 3G Igloo Lyo Stopper
OmniFlex 3G Serum Stopper
Ultra Pure
Ultra Pure Serum Stopper
Closure Color:
Blue
Red
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Closure Type Closure Color
06451222A 2 mL OmniFlex 3G Blue
06451225A 2 mL OmniFlex 3G Red
06451221A 2 mL OmniFlex 3G Blue
06451224A 2 mL OmniFlex 3G Red
06451220A 2 mL Ultra Pure Blue
06451223A 2 mL Ultra Pure Red
06451228A 5 mL OmniFlex 3G Igloo Lyo Stopper Blue
06451231A 5 mL OmniFlex 3G Igloo Lyo Stopper Red
06451227A 5 mL OmniFlex 3G Serum Stopper Blue
06451230A 5 mL OmniFlex 3G Serum Stopper Red
06451226A 5 mL Ultra Pure Serum Stopper Blue
06451229A 5 mL Ultra Pure Serum Stopper Red
06451234A 10 mL OmniFlex 3G Igloo Lyo Stopper Blue
06451237A 10 mL OmniFlex 3G Igloo Lyo Stopper Red
06451233A 10 mL OmniFlex 3G Serum Stopper Blue
06451236A 10 mL OmniFlex 3G Serum Stopper Red
06451232A 10 mL Ultra Pure Serum Stopper Blue
06451235A 10 mL Ultra Pure Serum Stopper Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
Catalog No. 06451222A Supplier DWK Life Sciences Supplier No. W991002CP
The WHEATON™ CompletePAK™ product line is a range of crimp top vials, rubber stoppers and seals that are sterilized ready-to-use and are off- the-shelf, and can be customized for any application reducing your supply chain due to our one source solution capability.
  • Sterilized, off-the-shelf, ready-to-use (RtU) products
  • Variety of Kitted and stand-alone sterile components – a solution for any application
  • Kitted solutions feature equal amounts of RtU components for reduced scrap rates
  • Complete manufacturing & certificate of analysis traceability
  • USP 788/EP 2.9.19 Particulate Matter in Injections, USP 85/EP 2.6.1, The Bacterial Endotoxins Test and USP 71/ EP 2.6.14 The Sterility Test, USP 660/EP 3.2.1 Glass Containers for Pharmaceutical Use, and USP 381/EP 3.2.9 Elastomeric Closures used in Parenteral Preparations
  • CompletePAK™ ModPAK Bulk configurations
  • Ready-to-Use, Just-in-Time, minimum 3 month shelf life from time of shipment
  • Container Closure Integrity (CCI) certification available upon request
Certifications
USP <788>, <85> and <71>; Container Closure Integrity (CCI) certified
Specifications
Closure Color Blue
Color Clear
Sterility Sterile
Volume (Metric) 2 mL
Includes Vial, Stopper, and Seal
No. per Case 278/Cs.
Product Line Wheaton CompletePAK
Closure Type OmniFlex 3G
Closure Size 13 mm
Certifications/Compliance USP 788, 85, and 71, Container Closure Integrity (CCI) certified