missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Clear Glass Sample Vials in Lab File with Caps Attached

Attached caps help maintain cleanliness. Wheaton™ Clear Glass Sample Vials in Lab File With Caps Attached come in partitioned trays to provide an easy way to store empty vials or to inventory samples.

$188.29 - $653.25

Specifications

Color Clear
Material Borosilicate Glass (Low Extractable)
For Use With (Application) Low profile for greater stability and less dead volume above the sample, making it ideal for compound storage
Thread Style Screw
Closure Material Phenolic
View More Specs

Products 17
Catalog Number Mfr. No. Volume (Metric) Closure Size Material (Liner) Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Closure Size Material (Liner) Price Quantity & Availability  
06408A
View Documents Promotion Details
DWK Life Sciences
224881
2 mL 8-425 14B Rubber
Case of 288 Each for $372.20
Only null left
 
06408B
View Documents Promotion Details
DWK Life Sciences
224882
4 mL 13-425 14B Rubber
Case of 144 Each for $199.16
Only null left
 
06408C
View Documents Promotion Details
DWK Life Sciences
224884
8 mL 15-425 14B Rubber
Case of 144 Each for $231.76
Only null left
 
0641211
View Documents Promotion Details
DWK Life Sciences
224885
12 mL 15-425 14B Rubber
Case of 144 Each for $253.61
Only null left
 
0641229
View Documents Promotion Details
DWK Life Sciences
225288
20 mL 24-400 14B Rubber
Case of 72 Each for $188.29
Only null left
 
0641230
View Documents Promotion Details
DWK Life Sciences
DWK225289
25 mL 24-400 14B Rubber
Case of 72 Each for $269.04
Only null left
 
0641231
View Documents Promotion Details
DWK Life Sciences
225290
40 mL 24-400 14B Rubber
Case of 72 Each for $356.68
Only null left
 
02911649
View Documents Promotion Details
DWK Life Sciences
W224582
4 mL 13-425 PTFE/14B Rubber
Case of 144 Each for $257.39
Only null left
 
02911648
View Documents Promotion Details
DWK Life Sciences
W224581
2 mL 8-425 PTFE/14B Rubber
Case of 288 Each for $653.25
Only null left
 
02911650
View Documents Promotion Details
DWK Life Sciences
224584
8 mL 15-425 PTFE/14B Rubber
Case of 144 Each for $291.76
Only null left
 
02911651
View Documents Promotion Details
DWK Life Sciences
W224585
12 mL 15-425 PTFE/14B Rubber
Case of 144 Each for $327.96
Only null left
 
02911652
View Documents Promotion Details
DWK Life Sciences
W224586
16 mL 18-400 PTFE/14B Rubber
Case of 144 Each for $470.14
Only null left
 
02911653
View Documents Promotion Details
DWK Life Sciences
W224588
24 mL 20-400 PTFE/14B Rubber
Case of 144 Each for $513.59
Only null left
 
02911665
View Documents Promotion Details
DWK Life Sciences
W224589
20 mL 24-400 PTFE/14B Rubber
Case of 72 Each for $375.96
Only null left
 
02911666
View Documents Promotion Details
DWK Life Sciences
W224590
25 mL 24-400 PTFE/14B Rubber
Case of 72 Each for $265.77
Only null left
 
02911667
View Documents Promotion Details
DWK Life Sciences
W224591
40 mL 24-400 PTFE/14B Rubber
Case of 72 Each for $474.39
Only null left
 
0333721A
View Documents Promotion Details
DWK Life Sciences
225012
4.0 mL 13-425 14B Rubber
Case of 144 Each for $247.43
Only null left
 
Description

Description

Attached caps help maintain cleanliness. Wheaton™ Clear Glass Sample Vials in Lab File With Caps Attached come in partitioned trays to provide an easy way to store empty vials or to inventory samples.

  • Fabricated from glass tubing for uniform sidewall and bottom thickness
  • Vials and caps are autoclavable
  • Clear vials made from low-extractable borosilicate glass that provides superior chemical resistance
  • Cap and Cap Liners
    • Solid top, black phenolic caps
    • 14B rubber-lined black phenolic screw cap for aqueous samples
    • PTFE/14B rubber-lined black phenolic screw cap for organic samples

Order Info

Replacements caps may be purchased separately.

Compliance

Clear vials conform to USP Type 1 and ASTM™ E 438 Type 1, Class A requirements.

Specifications

Specifications

Clear
Low profile for greater stability and less dead volume above the sample, making it ideal for compound storage
Phenolic
M-T File™ and Vial Rack
ASTM Type I, Class A, USP Type I
Borosilicate Glass (Low Extractable)
Screw
Screw Cap
Autoclavable
Videos
SDS
Documents

Documents