missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Clear Boston Rounds with Polyvinyl-lined Caps
Ideal for solvent, chemical and sample storage
$58.65 - $901.53
Specifications
| Color | Clear |
|---|---|
| Closure Color | White |
| Certifications/Compliance | EPA, FDA |
| Closure Liner | Poly/Vinyl |
| Closure Material | Polypropylene |
| Catalog Number | Mfr. No. | Capacity (Metric) | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Packaging | Price | Quantity | ||||
|
02-911-320
|
DWK Life Sciences
W216800 |
30 mL | Convenience pack |
Case of 48 Each for $143.50
|
|
||||
|
02-911-321
|
DWK Life Sciences
W216801 |
60 mL | Convenience pack |
Case of 24 Each for $85.97
|
|
||||
|
02-911-345
|
DWK Life Sciences
W216825 |
60 mL | Valu-bulk pack |
Case of 288 Each for $901.53
|
|
||||
|
02-911-346
|
DWK Life Sciences
W216826 |
125 mL | Valu-bulk pack |
Case of 160 Each for $439.97
|
|
||||
|
02-911-322
|
DWK Life Sciences
W216802 |
125 mL | Convenience pack |
Case of 24 Each for $96.18
|
|
||||
|
02-911-323
|
DWK Life Sciences
W216803 |
250 mL | Convenience pack |
Case of 12 Each for $58.65
|
|
||||
|
02-911-347
|
DWK Life Sciences
W216827 |
250 mL | Valu-bulk pack |
Case of 108 Each for $492.35
|
|
||||
|
02-911-324
|
DWK Life Sciences
W216804 |
500 mL | Convenience pack |
Case of 12 Each for $62.00
|
|
||||
|
02-911-325
|
DWK Life Sciences
W216805 |
1000 mL | Convenience pack |
Case of 12 Each for $108.21
|
|
||||
|
02-911-349
|
DWK Life Sciences
W216829 |
1000 mL | Valu-bulk pack |
Case of 12 Each for $118.71
|
|
||||
Description
Ideal for solvent, chemical and sample storage
- Narrow mouth enables liquids to be carefully poured to prevent spillage
- Convenience Pack bottles come with Hazard Communication labels for content identification, and Tamper Evident labels to protect integrity of samples in transit or long-term storage
- Available in partitioned Convenience or Valu-Bulk™ cartons
Order Info
Caps can be ordered separately.
Compliance
USP Type-III soda-lime glass
Specifications
| Clear | |
| EPA, FDA | |
| Polypropylene |
| White | |
| Poly/Vinyl | |
| Screw Cap |