missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Clear Boston Rounds with PE Cone-Lined Caps

Ideal for solvent, chemical and sample storage

Supplier:  DWK Life Sciences W216813

 View more versions of this product

Catalog No. 02-911-333

Only null left
Explore available promotions

Description

Description

  • Well suited for storage and sampling of liquids; clear Type III soda-lime glass provides visibility of contents
  • Narrow mouth enables liquids to be carefully poured to prevent spillage

Convenience Pack bottles come with Hazard Communication labels for content identification, and Tamper Evident labels to protect integrity of samples in transit or long-term storage.

Order Info

Caps can be ordered separately.

Specifications

Specifications

2 oz., 60 mL
2 oz.
1.53 in.
Black
94 mm
Polyethylene
20-400 mm
Screw Cap
60 mL
Clear
Soda Lime Glass
EPA, FDA
3.7 in.
Phenolic
39 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions