Learn More
DWK Life Sciences Wheaton™ Celstir™ Spinner Double Side Arm Flasks without Water Jacket
Catalog No. 064167B
Change view
Click to view available options
Capacity (Metric):
25 mL
50 mL
125 mL
250 mL
500 mL
1000 mL
3000 mL
Includes:
Flask only
Flask, impeller and caps
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Includes
064167B 50 mL Flask only
064162B 50 mL Flask, impeller and caps
064162D 250 mL Flask, impeller and caps
064162A 25 mL Flask, impeller and caps
064162F 3000 mL Flask, impeller and caps
064162J 1000 mL Flask, impeller and caps
064162C 125 mL Flask, impeller and caps
064162E 500 mL Flask, impeller and caps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 064167B Supplier DWK Life Sciences Supplier No. 356915
Provide maximum surface between culture and glass flask
  • PTFE-resin coated glass impeller does not protrude through cap and has bottom dimple for more efficient stirring
  • Silicone rubber-lined cap
  • Autoclavable
Specifications
Closure Material Silicone Rubber
Capacity (Metric) 50 mL
Material Borosilicate Glass
For Use With (Application) Suspension Culture
For Use With (Equipment) Magnetic Stirrer
Includes Flask only
Product Type Spinner Flask
Side Arm Joint 15-415
Side Arm Openings 2
Surface Coating PTFE Resin
Autoclavable Autoclavable
Graduated No
Sterility Non-sterile
Bottom Shape Flat
Closure Size 38-430
Quantity 1 Ea.
Show More Show Less