Learn More
DWK LIFE SCIENCES™ WHEATON™ CART2 Control, Monitor and Recording Software
Description
- Precisely controls rotating speeds of up to 255 separate roller units
- Each unit is remotely monitored for rotation, speed, deviation, and communication
- Records all settings, changes, alarms, and communication loss
- Password-protected user levels maintain security
- Can be easily added to R2P at any time
- Works with Windows-based software
- Provides a complete solution for GMP and FDA compliance requirements
Order Info
Computer requirements: IBM Pentium PC or compatible; 32 MB of RAM, minimum; Microsoft Windows 2000, XP; One free serial port; 10 MB of free hard drive space, minimum. Electrical requirements: 120VAC.
Compliance
Conforms to UL, CSA and CE Laboratory Standards
Specifications
Specifications
| Voltage | 120 V |
| Product Type | Monitor and recording software |
| For Use With (Equipment) | Wheaton R2P Roller Culture Apparatus |