missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Caps For Liquid Scintillation Vials

Catalog No. 0645090
Change view
Click to view available options
Material (Liner):
Metal Foil
Polyseal™
Material:
Urea
Closure Size:
15-425
22-400
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material (Liner) Material Closure Size
0645090 Polyseal™ Urea 22-400
0645091 Metal Foil Urea 15-425
0645092 Metal Foil Urea 22-400
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 0645090 Supplier DWK Life Sciences Supplier No. DWK240917
Only null left

Use as replacement caps or for vials that are provided without caps

Screw Caps For Liquid Scintillation Vials

  • Screw caps for WHEATON liquid scintillation vials
  • Case of 1000 provides an economical alterative
  • Not autoclavable

Specifications

Description Cap Size: 22-400
Material (Liner) Polyseal™
Color White
Material Urea
Product Line Wheaton
Quantity 1000 Cs.
Closure Size 22-400
For Use With (Equipment) Liquid Scintillation Vial
Autoclavable Not Autoclavable
Product Type Screw Cap