Learn More
DWK Life Sciences Wheaton™ Bottom-Drive R2P 2.0 Roller Apparatus
Advanced Production Scale Roller Culture Equipment
Supplier:  DWK Life Sciences WRBMR5061A
 View more versions of this product
Catalog No. 02924959
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description
Description
  • Color touch screen or digital display
  • Icon driven interface with intuitive menus
  • Surface mount gold plated electrical connections for better reliability in high temperature environments
  • Updated industry standard networking communications protocols and connections
  • Lower power consumption for longer battery run time and safer design
  • Backwards compatibility for continuing support of existing installations
  • Retrofit kits available to update older Wheaton systems or unsupported systems
  • Maintenance free brushless DC motors for years of trouble-free operation
Specifications
Chemical Identifiers
120 V
Bottles
For laboratory, pilot plant and production cell culture installations
Specifications
120 V
Bottom Drive Roller Culture System with options and 6 deck
30, Options
7 in.
Bottles
For laboratory, pilot plant and production cell culture installations
6/Pk.
Product Suggestions
Product Suggestions