Learn More
DWK Life Sciences Wheaton™ BOD Bottle Cap
Catalog No. 029267
Change view
Click to view available options
Bottle cap; For 60 and 300mL B.O.D. bottles
Catalog No.
029267 Bottle cap; For 60 and 300mL B.O.D. bottles
1 options
Catalog No. 029267 Supplier DWK Life Sciences Supplier No. 227723
For use with Wheaton 60 and 300mL BOD bottles and 300mL Black Borosilicate BOD bottles
  • Polyethylene cap prevents evaporation of water seal during incubation
Specifications
For Use With (Application) Prevents evaporation of the water seal during the five-day BOD incubation period
Material Polyethylene
For Use With (Equipment) For 60 and 300mL BOD bottle