missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Blue Starline Polypropylene Caps for Narrow-Mouth Containers

Catalog No. 0987714D
Change view
Click to view available options
Neck Finish:
20-410
24-410
28-410
38-430
No. per Case:
48 Cs.
72 Cs.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish No. per Case
0987714D 38-430 72 Cs.
0987711D 20-410 72 Cs.
0987712D 24-410 72 Cs.
0987713D 28-410 48 Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 0987714D Supplier DWK Life Sciences Supplier No. 23951604

Replacement caps for Leak-Resistant Containers

  • Leak-proof design
  • Autoclavable

Specifications

Color Blue
Material Polypropylene
No. per Case 72 Cs.
Neck Finish 38-430
Certifications/Compliance ISO 9001
For Use With (Equipment) Redi-pak™ leak resistant narrow mouth bottle