missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Wheaton™ Black Phenolic Screw Caps With PE Poly-Seal Liner
Description
- Poly-Seal liner is manufactured from low density polyethylene
- Screw Cap
- Phenolic
- Color: Black
It is recommended that liner be tested prior to use for leak resistant applications
Specifications
Specifications
| Color | Black |
| Material | Phenolic Resin |
| No. per Case | 144 Cs. |
| Neck Finish | 13-425 |
| For Use With (Equipment) | Bottle |
| Closure Liner | Polyethylene |
| Closure Type | Screw Cap |
| Certifications/Compliance | ISO 9001 |