missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Black Phenolic Screw Caps With PE Poly-Seal Liner

Catalog No. 02912526
Change view
Click to view available options
Neck Finish:
13-425
18-400
No. per Case:
144 Cs.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish No. per Case
02912526 18-400 144 Cs.
029843 13-425 144 Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 02912526 Supplier DWK Life Sciences Supplier No. 239251

Unique cone liner design offers superior torque retention and excellent sealing characteristics

  • Poly-Seal liner is manufactured from low density polyethylene
  • Screw Cap
  • Phenolic
  • Color: Black

It is recommended that liner be tested prior to use for leak resistant applications

Specifications

Color Black
Material Phenolic Resin
No. per Case 144 Cs.
Neck Finish 18-400
Certifications/Compliance ISO 9001
For Use With (Equipment) Bottle
Closure Liner Polyethylene
Closure Type Screw Cap