Learn More
DWK Life Sciences Wheaton™ Black Phenolic Screw Caps with PTFE Faced 14B White Rubber Liner
Catalog No. 03084126
Change view
Click to view available options
Closure Size:
24-400
8-425
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Size
03084126 24-400
03084117 8-425
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 03084126
Provides completely inert inner seal and surface for sample or product
  • Autoclavable
Specifications
Color Black
Material Phenolic
Liner Material TFE, Styrene Butadiene Rubber
Product Type Screw Cap
Closure Size 24-400
Autoclavable Autoclavable