Learn More
DWK Life Sciences Wheaton™ Black Phenolic Screw Caps with PTFE faced 14B Rubber Liners for E-Z Ex-Traction™ Vials
Designed to ensure the ultimate in product safety
Supplier: DWK Life Sciences W240821
Description
Designed to ensure the ultimate in product safety
- Autoclavable
- Liner made of styrene-butadiene rubber faced with PTFE
- Excellent chemical resistance
Chemical Identifiers
| Black | |
| Phenolic Resin | |
| ISO 9001 | |
| PTFE/Rubber |
Specifications
| Black | |
| 1000 Cs. | |
| ISO 9001 | |
| PTFE/Rubber |
| Phenolic Resin | |
| 13-425 | |
| 2 and 4mL Vial |