missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences Wheaton™ Black Phenolic Screw Cap with Metal Foil Liner

Features an aluminum foil faced pulp board liner that can be used with organic solvents

$369.04 - $369.04

Specifications

Color Black
Material Phenolic Resin
No. per Case 100 Cs.
Certifications/Compliance ISO 9001
For Use With (Equipment) Bottle
View More Specs

Products 1
Catalog Number Mfr. No. Neck Finish Price Quantity  
Catalog Number Mfr. No. Neck Finish Price Quantity  
06-450-37
View Documents
DWK Life Sciences
240321
33-400
Pack of 100 Each for $369.04
Only null left
 
Description

Description

  • Good resistance to hydrocarbons, oils, ketones and alcohols
  • Not good for acids or alkalis
  • Closure size: 33mm-400
  • Phenolic
  • Autoclavable
  • Black

Specifications

Specifications

Black
100 Cs.
Bottle
Phenolic Resin
ISO 9001
Metal Foil
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'