missing translation for 'onlineSavingsMsg'
Learn More

DWK LIFE SCIENCES™ WHEATON™ Aluminum Open Top Seals, Unlined
GREENER_CHOICE

Catalog No. 1112612
Change view
Click to view available options
Color:
Natural
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
1112612 Natural
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 1112612 Supplier DWK LIFE SCIENCES™ Supplier No. 22417801

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

WHEATON™ Aluminum Open Top Seals, Unlined expose the stopper for easy sample retrieval with a syringe. Available in a wide range of sizes and colors including natural, red, blue, and green, with good barrier properties and chemical resistance.

Open top seals expose stopper for sample retrieval with a syringe. Wheaton™ Aluminum Open Top Seals are available in a wide range of sizes and colors.

  • Open top design exposes stopper for sample retrieval with a syringe
  • Wide range of styles and sizes available
  • Color selection includes natural, red, blue, and green
  • Good barrier properties
  • Chemically resistant to alcohol and acetone, among others

Specifications

Color Natural
Diameter (Metric) Outer 20 mm
Material Aluminum
Pre-slit No
Sterility Non-sterile
Septum No
Quantity 1000/Cs.
Closure Size 20 mm
For Use With (Equipment) 20 mm Crimp Top Vials
Autoclavable Not Autoclavable
Product Type Open Top Crimp Cap
Show More Show Less