Learn More
DWK Life Sciences SOCOREX™ ACURA™ Macro Pipette Accessories, Nozzle Protection Filters
Description
- Add protection against liquid overfill or pipette contamination.
- Filters are interchangeable, non-sterile and non-autoclavable.
- Fits Socorex macro model pipettes
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Disposable | Single-use |
| Material | Polypropylene |
| Product Line | Acura |
| Sterility | Non-sterile |
| Volume (Metric) | 10 mL |
| For Use With (Equipment) | Socorex™ Macro Model Pipette |