missing translation for 'onlineSavingsMsg'
Learn More

DWK Life Sciences 520 Bottle-Top Dispenser Reservoirs

Catalog No. 137069
Change view
Click to view available options
Volume (Metric):
250 mL
500 mL
1000 mL
2500 mL
Color:
Amber
Diameter (Metric) Outer Neck:
32 mm
45 mm
Shape:
Round
Square
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Color Diameter (Metric) Outer Neck Shape
137069 500 mL Amber 32 mm Square
1370610 1000 mL Amber 45 mm Square
1370611 2500 mL Amber 45 mm Round
137067 250 mL Amber 32 mm Square
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 137069 Supplier DWK Life Sciences Supplier No. 844027

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Large selection of glass and Polyethylene reagent bottles, each supplied with one screw cap. Perfectly adapted with Calibrex™ bottle top dispensers.

  • Corresponding neck adapters supplied with the dispensers.

Specifications

Product Type Reagent Bottle
Color Amber
Material Soda Lime Glass
Volume (Metric) 500 mL
For Use With (Equipment) 520 model bottletop dispenser
Shape Square
Volume (English) 16.9 oz.
Diameter (English) Outer Neck 1.25 in.
Diameter (Metric) Outer Neck 32 mm