missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences 520 Bottle-Top Dispenser Reservoirs
Large selection of glass and Polyethylene reagent bottles, each supplied with one screw cap. Perfectly adapted with Calibrex™ bottle top dispensers.
Supplier: DWK Life Sciences 844026
Description
- Corresponding neck adapters supplied with the dispensers.
Specifications
| Reagent Bottle | |
| Borosilicate Glass | |
| 520 model bottletop dispenser | |
| 16.9 oz. | |
| 32 mm |
| Clear | |
| 500 mL | |
| Round | |
| 1.25 in. |