Learn More
DWK Life Sciences Wheaton™ 2D Data Matrix Bar Code Bottom Insert
For use with Wheaton CryoELITE™ Freestanding Cryogenic Vials
$136.75 - $512.19
Description
- Makes sample identification and traceability reliable and convenient
- Bar code insert manually press fits into bottom of vial
- Data matrix readable with standard scanning hardware
- Made from same cryogenic radiation-grade, virgin polypropylene as vial, ensuring uniform thermal expansion
Specifications
| Polypropylene | |
| 500/Cs. |