Learn More
DWK Life Sciences 12-Channel Modules for Acura Electro™ 956 Pipetters
For Acura Electro 956 12-Channel Pipetters
Supplier: DWK Life Sciences W870148
Description
- One control unit fits a choice of 27 volumetric assemblies available separately greatly extends working possibilities and makes electronic pipetting affordable to all budgets
- Disassembling and re-assembling without any tool contributes towards maximal flexibility
- Calibration specificities of extra modules, as set by factory QC or by pipette owner, are retained in instrument memory.
Specifications
| Rechargeable | |
| 956 | |
| 12-channel Module | |
| Ergonomic | |
| 2 Years |
| Universal Fit Pipette Tip | |
| 12 | |
| 20 to 350 μL | |
| 5 μL |