Learn More
Cytiva Whatman™ Reeve Angel 230 Creped Qualitative Circles
Provide reliable performance for non-critical industrial laboratory and student lab applications
Supplier:  Cytiva WHT5230185
Catalog No. 09-803E
Only null left
Explore available promotions
Description
Description
Retain coarse particles.
  • Made from cellulose
  • Creped surface for fast filtration of coarse particles
  • Particle retention: 25 to 30μm
  • Thickness: 0.40mm
Should not be used for kjeldahl nitrogen analysis
Product Suggestions
Product Suggestions