missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ pH Indicator, 1.0 to 11.0 Range, 10 books of 10 strips, pH Indicators and Test Papers
Description
Accurate for a wide range of routine pH testing
- Instant pH readings.
- Accurate for a wide range of routine pH testing.
- Inexpensive.
- Convenient and portable for field use.
Specifications
Specifications
| Color Indicator | Congo Red |
| pH Range | 1 to 11 |
| Format | Strip |
| For Use With (Application) | For the rapid determination of pH values has led to many applications in laboratories and industry |
| Graduations | 1 pH |
| Includes | 20 wide-range (pH 1 to 11) strips graduated in 1 pH units, color-comparison chart inside cover, 20 leaves per book, 10 sets of 10 books per pack |
| Type | pH Indicator Book |