missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Whatman™ Litmus Blue Test Paper

Catalog No. p-3207738
Change view
Click to view available options
Format:
Reel
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
0987577 Reel
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 0987577 Supplier Cytiva Supplier No. 2600201A
Only null left

Impregnated with a mixture of pH indicators, acid-alkali litmus papers assume a color specific to sample pH upon contact with a sample. Whatman™ Litmus Blue Test Paper provides rapid determination of pH values and has a variety of applications in laboratories and industry.

Mixed-indicator tests designed for fast, accurate, and reliable pH solution testing.
  • Instant pH readings
  • Accurate for a wide range of routine pH testing
  • Convenient and portable for field use
  • Color change in pH solutions ranging from approximately 5 to 8
  • Economical

Specifications

Color Indicator Blue
pH Range 5 to 8
Format Reel
Quantity 1 Each
Type Test Paper