missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva Whatman™ 96-Well Polypropylene UniPlate™ Microplates
Description
- One-piece molded collection and analysis plates
- Conforms to ANSI/SBS microplate standard
- Prevent optical cross talk in light emitting assays
- Untreated
- Ideal for filtrate collection, homogeneous assay techniques used in high-throughput screening (HTS)
Specifications
Specifications
| Format | Standard |
| Description | UNIPLATE microplates |
| Array | 8 x 12 |
| Color | Natural |
| Material | Polypropylene |
| Volume (Metric) Well | 2 mL |
| No. of Wells | 96 |
| Surface Treatment | Untreated |
| Well Shape | Square, Round Bottom (U-well) |
| For Use With (Application) | For homogeneous assays techniques utilized in high-throughput screening |