missing translation for 'onlineSavingsMsg'
Learn More

What Leaf Is It?

Catalog No. S19555
Change view
Click to view available options
Grade:
3 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S19555 3 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S19555 Supplier Customer Channels Group (Ccg) Supplier No. 201 EMD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

  • Two each of 13 identified leaves
  • 26 unidentified leaves
  • Maple, oak, elm, ash and other varieties of leaves carded, labeled and bagged for protection
  • Leaf key to identify unknown leaf specimens
  • Picture glossary of leaf terms
  • 61-page Tree Finder book
  • Teacher's guide with hands-on activities

Use actual leaf specimens to identify trees.

A fun and economical way to learn how to key out, identify and classify real leaves by their shape, size, veins, margins and position on a twig.

Specifications

Grade 3 to 12
For Use With (Application) Students key out, identify and classify real leaves by their shape, size, veins, margins and position on twig
Includes 39 Real tree leaves (each leaf is carded, labeled and bagged), maple, oak, elm, ash and other varieties of leaves carded, labeled and bagged for protection, leaf key to identify unknown leaf specimens, picture glossary of leaf terms, 61-page tree finder book, teacher's guide with hands-on activities
Product Type Leaf Identification Kit