Research Products Corporation WetGrip Micro-Tube Laser Printer Labels, Fits 0.5ml Tubes, 0.94 x 0.5 Inches, 2380 per Package
Catalog No. 50998653
Ideal for ice baths, WetGrip Micro-Tube Labels grip to wet or frozen tubes and microscope slides. Labels adhere to glass, plastic, metal and paper. Recommended temperature range is -80°C to 100°C. Supplied on laser printable sheets for printing many labels at once.
Catalog No.
50998653
Supplier
Research Products CorporationSupplier No.
247176
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Ideal for ice baths, WetGrip Micro-Tube Labels grip to wet or frozen tubes and microscope slides. Labels adhere to glass, plastic, metal and paper. Recommended temperature range is -80°C to 100°C. Supplied on laser printable sheets for printing many labels at once.