missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ WesternDot™ 625 Western Blot Kits
Description
The WesternDot 625 Western Blot kits combine the unique properties of Qdot 625 nanocrystals with the high-affinity streptavidin-biotin interaction to allow simple, highly sensitive detection of proteins on western blots. The WesternDot kits contain optimized, ready-to-use or ready-to-dilute reagents for sensitive immunodetection of proteins immobilized on nitrocellulose (NC) or polyvinylidene difluoride (PVDF) membranes.
WesternDot 625 Western Blot kits utilize extremely bright Qdot 625 nanocrystals and include reagents for sensitive imaging of proteins probed with mouse or rabbit primary antibodies.
Features include:
• Detection performance and ease-of-use exceed existing ECL-based methods
• Make use of extremely bright Qdot 625 streptavidin conjugate to allow direct imaging of blots while avoiding the need to spend time and money on optimizing multiple exposures to film in the dark
• Contains reagents sufficient for 20 mini-gel blots (7 x 10 cm/blot)
Features include:
• Detection performance and ease-of-use exceed existing ECL-based methods
• Make use of extremely bright Qdot 625 streptavidin conjugate to allow direct imaging of blots while avoiding the need to spend time and money on optimizing multiple exposures to film in the dark
• Contains reagents sufficient for 20 mini-gel blots (7 x 10 cm/blot)
Order Info
Shipping Condition: Room Temperature
Specifications
Specifications
| Content And Storage | • WesternDot™ Blocking Buffer, Component A • Wash Buffer, Component B • Biotin-XX Goat Anti-Mouse IgG (H+L), Component C • Qdot™ 625 Streptavidin Conjugate, Component D • WesternDot™ Staining Dish, Component E Store at 4°C, Do Not Freeze. |
| Detection Location | In-Blot Detection |
| Color | Red |
| Detection Method | Fluorescence |
| Label or Dye | Qdot 625 |
| Label Type | Qdot Nanocrystals |
| Quantity | 1 kit |
| Reactivity | Mouse |
| Sample Type | Protein, Cell Extracts |
| Shipping Condition | Room Temperature |
| Show More |