This item has a minimum qty of 2 per supplier requirements. MultiBox™, 2 compartments, 85 x 85 x 30mm each (311/32 x 311/32 x 13/16 in.), for Novex Mini (8x8cm), Bio-Rad Mini (7x8cm) and others
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
This item has a minimum qty of 2 per supplier requirements. MultiBox™, 2 compartments, 85 x 85 x 30mm each (311/32 x 311/32 x 13/16 in.), for Novex Mini (8x8cm), Bio-Rad Mini (7x8cm) and others