Learn More
Electron Microscopy Sciences Western Blotting Boxes Square, Clear, 7 ml, 4-9/16x 4-9/16 x 1""
Catalog No. 501929216
Encompass
No matter what size your Western blot is, there's a box that fits it perfectly. With boxes that are correctly matched to common blot sizes, you can fit more blots on shakers/rockers while keeping reagent and antibody usage to an absolute minimum. Additionally, all corners are formed at a 90 deg angle and the bottom surface is uniformly flat, further reducing consumption of costly reagents and antibodies. Equipped with an easy-to-open hinged lid trhat reduces evaporation. Applications: Owl Penguin P9DS/Puffin.
Color: Clear
Max volume: 7 ml
Dimensions: 4-9/16x 4-9/16 x 1"
Catalog No. 501929216 Supplier Electron Microscopy Sciences Supplier No. 7047509
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
No matter what size your Western blot is, there's a box that fits it perfectly. With boxes that are correctly matched to common blot sizes, you can fit more blots on shakers/rockers while keeping reagent and antibody usage to an absolute minimum. Additionally, all corners are formed at a 90 deg angle and the bottom surface is uniformly flat, further reducing consumption of costly reagents and antibodies. Equipped with an easy-to-open hinged lid trhat reduces evaporation. Applications: Owl Penguin P9DS/Puffin.
Color: Clear
Max volume: 7 ml
Dimensions: 4-9/16x 4-9/16 x 1"