Li Cor Biotech Llc Black Western Blot Incubation Boxes (medium - 10 boxes)
Catalog No. NC9358182
Black blot boxes are suitable for incubating near-infrared fluorescent and chemiluminescent Western blots or protein gels. The Medium size is: 8.9 cm x 6.5 cm x 2.8 cm.
Catalog No.
NC9358182
Supplier
Li Cor Biotech LlcSupplier No.
92997210
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Black blot boxes are suitable for incubating near-infrared fluorescent and chemiluminescent Western blots or protein gels. The Medium size is: 8.9 cm x 6.5 cm x 2.8 cm.